Assay Details
| Gene Symbol: ddt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002147.1 | NP_001002147.1 | 2 - 3 | 323 | 80 | |
| GenBank mRNA | BC071391.1 | - | 2 - 3 | 323 | 80 | |
More Information
Set Membership: |
|

ddt
Genomic Map