Assay Details
| Gene Symbol: blvrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002686.1 | NP_001002686.1 | 4 - 5 | 595 | 110 | |
| GenBank mRNA | BC076451.1 | - | 4 - 5 | 595 | 110 | |
| BC164991.1 | - | 4 - 5 | 458 | 110 | ||
More Information
Set Membership: |
|

blvrb
Genomic Map