Assay Details
| Gene Symbol: ttc33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002722.1 | NP_001002722.1 | 2 - 3 | 308 | 59 | |
| GenBank mRNA | BC075750.1 | - | 2 - 3 | 308 | 59 | |
More Information
Set Membership: |
|

ttc33
Genomic Map