Assay Details
Gene Symbol: parvaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002872.2 | NP_001002872.1 | 2 - 3 | 353 | 82 | |
GenBank mRNA | AY648735.1 | - | 2 - 3 | 224 | 82 | |
BC085399.1 | - | 2 - 3 | 353 | 82 | ||
BC164207.1 | - | 2 - 3 | 258 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
