Assay Details
Gene Symbol: rs1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003438.2 | NP_001003438.1 | 1 - 2 | 64 | 72 | |
GenBank mRNA | BC076183.1 | - | 1 - 2 | 65 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
