Assay Details
Gene Symbol: rs1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003438.2 | NP_001003438.1 | 3 - 4 | 216 | 80 | |
GenBank mRNA | BC076183.1 | - | 3 - 4 | 217 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
