Assay Details
Gene Symbol: lsm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003551.1 | NP_001003551.1 | 3 - 4 | 323 | 109 | |
GenBank mRNA | BC078334.1 | - | 3 - 4 | 323 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
