Assay Details
| Gene Symbol: naa50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003623.1 | NP_001003623.1 | 2 - 3 | 262 | 89 | |
| XM_005162691.4 | XP_005162748.1 | 2 - 3 | 89 | |||
| GenBank mRNA | BC078316.1 | - | 2 - 3 | 262 | 89 | |
More Information
Set Membership: |
|

naa50
Genomic Map