Assay Details
Gene Symbol: naa50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003623.1 | NP_001003623.1 | 3 - 4 | 383 | 80 | |
XM_005162691.4 | XP_005162748.1 | 3 - 4 | 423 | 80 | ||
GenBank mRNA | BC078316.1 | - | 3 - 4 | 383 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
