Assay Details
| Gene Symbol: sulf2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003833.2 | NP_001003833.2 | 14 - 15 | 2255 | 95 | |
| XM_017353509.1 | XP_017208998.1 | 14 - 15 | 2265 | 95 | ||
| XM_017353510.2 | XP_017208999.1 | 16 - 17 | 2568 | 95 | ||
| XM_017353511.2 | XP_017209000.1 | 14 - 15 | 2169 | 95 | ||
| XM_021469735.1 | XP_021325410.1 | 18 - 19 | 2987 | 95 | ||
| XM_021469736.1 | XP_021325411.1 | 13 - 14 | 2035 | 95 | ||
| XM_021469737.1 | XP_021325412.1 | 13 - 14 | 2035 | 95 | ||
| XM_021469738.1 | XP_021325413.1 | 13 - 14 | 2035 | 95 | ||
| XM_021469739.1 | XP_021325414.1 | 13 - 14 | 2035 | 95 | ||
| GenBank mRNA | AY332607.1 | - | 14 - 15 | 2255 | 95 | |
| AY332608.1 | - | 13 - 14 | 2035 | 95 | ||
More Information
Set Membership: |
|

sulf2b
Genomic Map