Assay Details
| Gene Symbol: sulf2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003833.2 | NP_001003833.2 | 18 - 19 | 2757 | 92 | |
| XM_021469736.1 | XP_021325411.1 | 17 - 18 | 2537 | 92 | ||
| GenBank mRNA | AY332607.1 | - | 18 - 19 | 2757 | 92 | |
More Information
Set Membership: |
|

sulf2b
Genomic Map