Assay Details
| Gene Symbol: sulf2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003833.2 | NP_001003833.2 | 12 - 13 | 2065 | 64 | |
| XM_017353509.1 | XP_017208998.1 | 12 - 13 | 2075 | 64 | ||
| XM_017353510.2 | XP_017208999.1 | 14 - 15 | 2378 | 64 | ||
| XM_017353511.2 | XP_017209000.1 | 12 - 13 | 1979 | 64 | ||
| XM_021469735.1 | XP_021325410.1 | 16 - 17 | 2797 | 64 | ||
| XM_021469736.1 | XP_021325411.1 | 11 - 12 | 1845 | 64 | ||
| XM_021469737.1 | XP_021325412.1 | 11 - 12 | 1845 | 64 | ||
| XM_021469738.1 | XP_021325413.1 | 11 - 12 | 1845 | 64 | ||
| XM_021469739.1 | XP_021325414.1 | 11 - 12 | 1845 | 64 | ||
| GenBank mRNA | AY332607.1 | - | 12 - 13 | 2065 | 64 | |
| AY332608.1 | - | 11 - 12 | 1845 | 64 | ||
More Information
Set Membership: |
|

sulf2b
Genomic Map