Assay Details
Gene Symbol: ebna1bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003840.1 | NP_001003840.1 | 4 - 5 | 514 | 74 | |
GenBank mRNA | AY648829.1 | - | 4 - 5 | 514 | 74 | |
BC153551.1 | - | 4 - 5 | 484 | 74 | ||
BC162284.1 | - | 4 - 5 | 512 | 74 | ||
BC162298.1 | - | 4 - 5 | 512 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
