Assay Details
| Gene Symbol: spic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004621.2 | NP_001004621.1 | 4 - 5 | 430 | 133 | |
| GenBank mRNA | BC081405.1 | - | 4 - 5 | 431 | 133 | |
| BC164951.1 | - | 3 - 4 | 348 | 133 | ||
More Information
Set Membership: |
|

spic
Genomic Map