Assay Details
| Gene Symbol: pgrmc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007392.1 | NP_001007393.1 | 1 - 2 | 300 | 81 | |
| GenBank mRNA | BC085558.1 | - | 1 - 2 | 300 | 81 | |
| BC164955.1 | - | 1 - 2 | 325 | 81 | ||
More Information
Set Membership: |
|

pgrmc1
Genomic Map