Assay Details
| Gene Symbol: gpr137c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007433.1 | NP_001007434.1 | 3 - 4 | 958 | 58 | |
| XM_005169819.4 | XP_005169876.1 | 3 - 4 | 58 | |||
| GenBank mRNA | BC085410.1 | - | 3 - 4 | 958 | 58 | |
More Information
Set Membership: |
|

gpr137c
Genomic Map