Assay Details
| Gene Symbol: kat5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013309.3 | NP_001013327.2 | 11 - 12 | 1205 | 70 | |
| XM_021476135.1 | XP_021331810.1 | 11 - 12 | 1339 | 70 | ||
| XM_021476136.1 | XP_021331811.1 | 10 - 11 | 1270 | 70 | ||
| GenBank mRNA | BC090743.1 | - | 11 - 12 | 1174 | 70 | |
More Information
Set Membership: |
|

kat5b
Genomic Map