Assay Details
| Gene Symbol: kat5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013309.3 | NP_001013327.2 | 7 - 8 | 653 | 125 | |
| XM_021476135.1 | XP_021331810.1 | 7 - 8 | 736 | 125 | ||
| XM_021476136.1 | XP_021331811.1 | 6 - 7 | 667 | 125 | ||
| GenBank mRNA | BC090743.1 | - | 7 - 8 | 622 | 125 | |
More Information
Set Membership: |
|

kat5b
Genomic Map