Assay Details
| Gene Symbol: kat5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013309.3 | NP_001013327.2 | 9 - 10 | 963 | 127 | |
| GenBank mRNA | BC090743.1 | - | 9 - 10 | 932 | 127 | |
More Information
Set Membership: |
|

kat5b
Genomic Map