Assay Details
Gene Symbol: pcyt1aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017634.1 | NP_001017634.1 | 4 - 5 | 423 | 118 | |
XM_021480218.1 | XP_021335893.1 | 5 - 6 | 643 | 118 | ||
GenBank mRNA | BC092816.1 | - | 4 - 5 | 423 | 118 | |
BC164470.1 | - | 3 - 4 | 358 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
