Assay Details
Gene Symbol: pcyt1aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017634.1 | NP_001017634.1 | 6 - 7 | 655 | 71 | |
XM_021480218.1 | XP_021335893.1 | 7 - 8 | 875 | 71 | ||
GenBank mRNA | BC092816.1 | - | 6 - 7 | 655 | 71 | |
BC164470.1 | - | 5 - 6 | 590 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
