Assay Details
| Gene Symbol: rab3ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017761.1 | NP_001017761.1 | 4 - 5 | 537 | 75 | |
| GenBank mRNA | BC093355.1 | - | 4 - 5 | 537 | 75 | |
| BC164318.1 | - | 3 - 4 | 503 | 75 | ||
More Information
Set Membership: |
|

rab3ab
Genomic Map