Assay Details
| Gene Symbol: ogt.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017359.1 | NP_001017359.1 | 18 - 19 | 2431 | 66 | |
| NM_001018105.1 | NP_001018115.1 | 18 - 19 | 2401 | 66 | ||
| XM_009307793.2 | XP_009306068.1 | 15 - 16 | 3782 | 66 | ||
| GenBank mRNA | AY696178.1 | - | 18 - 19 | 2431 | 66 | |
| AY696179.1 | - | 18 - 19 | 2401 | 66 | ||
More Information
Set Membership: |
|

ogt.1
Genomic Map