Assay Details
| Gene Symbol: dnmt3ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020476.2 | NP_001018312.2 | 11 - 12 | 2854 | 95 | |
| XM_005166992.4 | XP_005167049.1 | 12 - 13 | 3070 | 95 | ||
| XM_005166993.4 | XP_005167050.1 | 12 - 13 | 3043 | 95 | ||
| XM_005166994.4 | XP_005167051.1 | 11 - 12 | 3001 | 95 | ||
| XM_005166996.4 | XP_005167053.1 | 10 - 11 | 2779 | 95 | ||
| GenBank mRNA | AB196918.1 | - | 10 - 11 | 2785 | 95 | |
| BC163546.1 | - | 11 - 12 | 2846 | 95 | ||
More Information
Set Membership: |
|

dnmt3ba
Genomic Map