Assay Details
| Gene Symbol: dnmt3ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020476.2 | NP_001018312.2 | 17 - 18 | 3528 | 89 | |
| XM_005166992.4 | XP_005167049.1 | 18 - 19 | 3744 | 89 | ||
| XM_005166993.4 | XP_005167050.1 | 18 - 19 | 3717 | 89 | ||
| XM_005166994.4 | XP_005167051.1 | 17 - 18 | 3675 | 89 | ||
| XM_005166996.4 | XP_005167053.1 | 16 - 17 | 3453 | 89 | ||
| GenBank mRNA | AB196918.1 | - | 16 - 17 | 3459 | 89 | |
More Information
Set Membership: |
|

dnmt3ba
Genomic Map