Assay Details
| Gene Symbol: dnmt3ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020476.2 | NP_001018312.2 | 19 - 20 | 3760 | 63 | |
| XM_005166992.4 | XP_005167049.1 | 20 - 21 | 3976 | 63 | ||
| XM_005166993.4 | XP_005167050.1 | 20 - 21 | 3949 | 63 | ||
| XM_005166994.4 | XP_005167051.1 | 19 - 20 | 3907 | 63 | ||
| XM_005166996.4 | XP_005167053.1 | 18 - 19 | 3685 | 63 | ||
| GenBank mRNA | AB196918.1 | - | 18 - 19 | 3691 | 63 | |
| BC163546.1 | - | 19 - 20 | 3752 | 63 | ||
More Information
Set Membership: |
|

dnmt3ba
Genomic Map