Assay Details
| Gene Symbol: dnmt3ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020476.2 | NP_001018312.2 | 20 - 21 | 3848 | 90 | |
| XM_005166992.4 | XP_005167049.1 | 21 - 22 | 4064 | 90 | ||
| XM_005166993.4 | XP_005167050.1 | 21 - 22 | 4037 | 90 | ||
| XM_005166994.4 | XP_005167051.1 | 20 - 21 | 3995 | 90 | ||
| XM_005166996.4 | XP_005167053.1 | 19 - 20 | 3773 | 90 | ||
| GenBank mRNA | AB196918.1 | - | 19 - 20 | 3779 | 90 | |
| BC163546.1 | - | 20 - 21 | 3840 | 90 | ||
More Information
Set Membership: |
|

dnmt3ba
Genomic Map