Assay Details
| Gene Symbol: dnmt3ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020476.2 | NP_001018312.2 | 2 - 3 | 167 | 122 | |
| XM_005166992.4 | XP_005167049.1 | 2 - 3 | 251 | 122 | ||
| XM_005166993.4 | XP_005167050.1 | 2 - 3 | 224 | 122 | ||
| XM_005166994.4 | XP_005167051.1 | 2 - 3 | 251 | 122 | ||
| XM_005166996.4 | XP_005167053.1 | 2 - 3 | 161 | 122 | ||
| GenBank mRNA | AB196918.1 | - | 2 - 3 | 167 | 122 | |
| BC163546.1 | - | 2 - 3 | 159 | 122 | ||
More Information
Set Membership: |
|

dnmt3ba
Genomic Map