Assay Details
| Gene Symbol: dnmt3ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020476.2 | NP_001018312.2 | 8 - 9 | 2536 | 69 | |
| XM_005166996.4 | XP_005167053.1 | 7 - 8 | 2461 | 69 | ||
| GenBank mRNA | AB196918.1 | - | 7 - 8 | 2467 | 69 | |
More Information
Set Membership: |
|

dnmt3ba
Genomic Map