Assay Details
| Gene Symbol: pcyt1ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020735.1 | NP_001018571.1 | 5 - 6 | 644 | 65 | |
| GenBank mRNA | BC095130.1 | - | 5 - 6 | 644 | 65 | |
More Information
Set Membership: |
|

pcyt1ab
Genomic Map