Assay Details
| Gene Symbol: cox5aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024403.2 | NP_001019574.1 | 2 - 3 | 278 | 76 | |
| GenBank mRNA | BC095110.1 | - | 2 - 3 | 219 | 76 | |
| BC097215.1 | - | 2 - 3 | 219 | 76 | ||
| BC164277.1 | - | 2 - 3 | 218 | 76 | ||
| EH560056.1 | - | 3 - 4 | 434 | 76 | ||
| EL647681.1 | - | 2 - 3 | 291 | 76 | ||
More Information
Set Membership: |
|

cox5aa
Genomic Map