Assay Details
| Gene Symbol: cox5aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024403.2 | NP_001019574.1 | 4 - 5 | 537 | 72 | |
| GenBank mRNA | BC095110.1 | - | 4 - 5 | 478 | 72 | |
| BC097215.1 | - | 4 - 5 | 478 | 72 | ||
| EH560056.1 | - | 1 - 2 | 175 | 72 | ||
More Information
Set Membership: |
|

cox5aa
Genomic Map