Assay Details
| Gene Symbol: nedd4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034186.1 | NP_001029358.1 | 13 - 14 | 1142 | 116 | |
| GenBank mRNA | AY995154.1 | - | 13 - 14 | 1142 | 116 | |
More Information
Set Membership: |
|

nedd4a
Genomic Map