Assay Details
| Gene Symbol: nedd4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034186.1 | NP_001029358.1 | 17 - 18 | 1576 | 101 | |
| GenBank mRNA | AY995154.1 | - | 17 - 18 | 1576 | 101 | |
More Information
Set Membership: |
|

nedd4a
Genomic Map