Assay Details
| Gene Symbol: si:dkey-102c8.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044345.1 | NP_001037810.1 | 3 - 4 | 910 | 102 | |
| XM_005173958.4 | XP_005174015.1 | 4 - 5 | 855 | 102 | ||
| XM_009296103.3 | XP_009294378.1 | 4 - 5 | 870 | 102 | ||
| GenBank mRNA | BC135063.1 | - | 3 - 4 | 910 | 102 | |
| Gene Symbol: sc:d0220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115107.1 | NP_001108579.1 | 3 - 4 | 1005 | 102 | |
| GenBank mRNA | CU458966.1 | - | 3 - 4 | 1005 | 102 | |
| Gene Symbol: si:dkey-102c8.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BM956921.1 | - | 1 - 2 | 122 | 102 | |
More Information
Set Membership: |
|

si:dkey-102c8.3
Genomic Map