Assay Details
Gene Symbol: taf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044395.2 | NP_001037860.2 | 1 - 2 | 151 | 73 | |
GenBank mRNA | BC076279.1 | - | 1 - 2 | 71 | 73 | |
BC122276.1 | - | 1 - 2 | 118 | 73 | ||
BC153648.1 | - | 1 - 2 | 71 | 73 | ||
BC165107.1 | - | 1 - 2 | 68 | 73 | ||
EH498335.1 | - | 1 - 2 | 180 | 73 | ||
EH555285.1 | - | 1 - 2 | 123 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
