Assay Details
Gene Symbol: taf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044395.2 | NP_001037860.2 | 2 - 3 | 228 | 84 | |
GenBank mRNA | BC076279.1 | - | 2 - 3 | 148 | 84 | |
BC095147.1 | - | 2 - 3 | 137 | 84 | ||
BC122276.1 | - | 2 - 3 | 195 | 84 | ||
BC153648.1 | - | 2 - 3 | 148 | 84 | ||
BC165107.1 | - | 2 - 3 | 145 | 84 | ||
EH498335.1 | - | 2 - 3 | 257 | 84 | ||
EH555285.1 | - | 2 - 3 | 200 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
