Assay Details
Gene Symbol: taf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044395.2 | NP_001037860.2 | 6 - 7 | 698 | 79 | |
GenBank mRNA | BC076279.1 | - | 6 - 7 | 618 | 79 | |
BC095147.1 | - | 6 - 7 | 607 | 79 | ||
BC122276.1 | - | 6 - 7 | 665 | 79 | ||
BC153648.1 | - | 6 - 7 | 618 | 79 | ||
BC165107.1 | - | 6 - 7 | 615 | 79 | ||
EH498335.1 | - | 6 - 7 | 727 | 79 | ||
EH555285.1 | - | 6 - 7 | 670 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
