Assay Details
Gene Symbol: poc1bl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044778.2 | NP_001038243.2 | 4 - 5 | 1040 | 122 | |
GenBank mRNA | BC090692.1 | - | 4 - 5 | 1040 | 122 | |
BC164662.1 | - | 3 - 4 | 777 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
