Assay Details
Gene Symbol: poc1bl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044778.2 | NP_001038243.2 | 6 - 7 | 1341 | 101 | |
GenBank mRNA | BC090692.1 | - | 6 - 7 | 1341 | 101 | |
BC164662.1 | - | 5 - 6 | 1078 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
