Assay Details
Gene Symbol: poc1bl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044778.2 | NP_001038243.2 | 8 - 9 | 1649 | 86 | |
GenBank mRNA | BC090692.1 | - | 8 - 9 | 1649 | 86 | |
BC164662.1 | - | 7 - 8 | 1386 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
