Assay Details
| Gene Symbol: nars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077155.1 | NP_001070623.1 | 2 - 3 | 489 | 56 | |
| XM_021481436.1 | XP_021337111.1 | 2 - 3 | 526 | 56 | ||
| GenBank mRNA | BC124277.1 | - | 2 - 3 | 489 | 56 | |
More Information
Set Membership: |
|

nars2
Genomic Map