Assay Details
| Gene Symbol: mrpl21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077468.1 | NP_001070936.1 | 1 - 2 | 143 | 74 | |
| XM_005172382.4 | XP_005172439.1 | 1 - 2 | 133 | 74 | ||
| GenBank mRNA | BC124336.1 | - | 1 - 2 | 143 | 74 | |
More Information
Set Membership: |
|

mrpl21
Genomic Map