Assay Details
| Gene Symbol: atp2a1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077533.1 | NP_001071001.1 | 12 - 13 | 1561 | 93 | |
| XM_005156072.4 | XP_005156129.1 | 12 - 13 | 1595 | 93 | ||
| GenBank mRNA | BC051779.1 | - | 13 - 14 | 1445 | 93 | |
| BC125955.1 | - | 12 - 13 | 1561 | 93 | ||
More Information
Set Membership: |
|

atp2a1l
Genomic Map