Assay Details
| Gene Symbol: atp2a1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077533.1 | NP_001071001.1 | 23 - 24 | 3148 | 87 | |
| GenBank mRNA | BC051779.1 | - | 24 - 25 | 2817 | 87 | |
| BC125955.1 | - | 23 - 24 | 3148 | 87 | ||
More Information
Set Membership: |
|

atp2a1l
Genomic Map