Assay Details
Gene Symbol: atp2a1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077533.1 | NP_001071001.1 | 5 - 6 | 598 | 61 | |
XM_005156072.4 | XP_005156129.1 | 5 - 6 | 632 | 61 | ||
GenBank mRNA | BC051779.1 | - | 6 - 7 | 482 | 61 | |
BC125955.1 | - | 5 - 6 | 598 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
