Assay Details
Gene Symbol: stxbp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089376.1 | NP_001082845.1 | 3 - 4 | 210 | 82 | |
GenBank mRNA | BC171526.1 | - | 3 - 4 | 192 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
