Assay Details
Gene Symbol: stxbp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089376.1 | NP_001082845.1 | 4 - 5 | 288 | 78 | |
GenBank mRNA | BC134211.1 | - | 4 - 5 | 281 | 78 | |
BC171526.1 | - | 4 - 5 | 270 | 78 | ||
BC171552.1 | - | 4 - 5 | 270 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
