Assay Details
| Gene Symbol: anapc11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098480.1 | NP_001091950.1 | 2 - 3 | 171 | 93 | |
| XM_005166196.4 | XP_005166253.1 | 2 - 3 | 198 | 93 | ||
| GenBank mRNA | BC124716.1 | - | 2 - 3 | 171 | 93 | |
More Information
Set Membership: |
|

anapc11
Genomic Map