Assay Details
| Gene Symbol: cyp2aa7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109722.1 | NP_001103192.1 | 1 - 2 | 256 | 82 | |
| GenBank mRNA | BC152008.1 | - | 1 - 2 | 256 | 82 | |
| Gene Symbol: cyp2aa11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017353668.2 | XP_017209157.1 | 1 - 2 | 251 | 82 | |
More Information
Set Membership: |
|

cyp2aa7
Genomic Map